Learn More
Biomatik Corporation Rat Prolactin Induced Protein (PIP) ELISA Kit
Supplier:  Biomatik Corporation EKU06826
Encompass_Preferred
Rat Prolactin Induced Protein (PIP) ELISA Kit for detecting PIP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.88-3000pg/ml. Sensitivity: 16.23pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70417. Gene ID: 64673. Target Synonyms: GCDFP15; GPIP4; BRST2; SABP; gp17; Gross cystic disease fluid protein 15; Prolactin-induced protein; Secretory actin-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8806