Learn More
Biomatik Corporation Rat Prolylcarboxypeptidase (PRCP) ELISA Kit
Supplier:  Biomatik Corporation EKU06836
Encompass_Preferred
Rat Prolylcarboxypeptidase (PRCP) ELISA Kit for detecting PRCP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4AA31. Gene ID: 293118. Target Synonyms: PCP; HUMPCP; Lysosomal Pro-X Carboxypeptidase; Angiotensinase C; Lysosomal carboxypeptidase C; Proline carboxypeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5675