Learn More
Biomatik Corporation Rat Proopiomelanocortin (POMC) ELISA Kit
Supplier:  Biomatik Corporation EKU11694
Encompass_Preferred
Rat Proopiomelanocortin (POMC) ELISA Kit for detecting POMC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01194. Target Synonyms: POC; ProOMC; Pro-OMC; Pro Opiomelanocortin; Corticotropin-lipotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4866