Learn More
Biomatik Corporation Rat Protease Activated Receptor 1 (PAR1) ELISA Kit
Supplier:  Biomatik Corporation EKU06870
Encompass_Preferred
Rat Protease Activated Receptor 1 (PAR1) ELISA Kit for detecting PAR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26824. Target Synonyms: FIIR; F2r; CF2R; HTR; PAR1; TR; Coagulation Factor II Receptor; Thrombin Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6973