Learn More
Biomatik Corporation Rat Protease Activated Receptor 4 (PAR4) ELISA Kit
Supplier:  Biomatik Corporation EKU06875
Encompass_Preferred
Rat Protease Activated Receptor 4 (PAR4) ELISA Kit for detecting PAR4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q920E0. Gene ID: 116498. Target Synonyms: F2RL3; PAR-4; Coagulation Factor II Thrombin Receptor-Like 3; Coagulation factor II receptor-like 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6997