Learn More
Biomatik Corporation Rat Protein Kinase C Beta 1 (PKCb1) ELISA Kit
Supplier:  Biomatik Corporation EKU06918
Encompass_Preferred
Rat Protein Kinase C Beta 1 (PKCb1) ELISA Kit for detecting PKCb1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P68403. Gene ID: 25023. Target Synonyms: PKC-B1; PRKCB; PRKCB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4890