Learn More
Biomatik Corporation Rat Protein Kinase C Gamma (PKCg) ELISA Kit
Supplier:  Biomatik Corporation EKU11376
Encompass_Preferred
Rat Protein Kinase C Gamma (PKCg) ELISA Kit for detecting PKCg in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63319. Gene ID: 24681. Target Synonyms: PKC-G; PRKCG; PKCC; SCA14; PKC-Gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7798