Learn More
Biomatik Corporation Rat Protein Kinase, X-Linked (PRKX) ELISA Kit
Supplier:  Biomatik Corporation EKU06935
Encompass_Preferred
Rat Protein Kinase, X-Linked (PRKX) ELISA Kit for detecting PRKX in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5BK52. Gene ID: 501563. Target Synonyms: PKX1; cAMP-dependent protein kinase catalytic subunit PRKX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8796