Learn More
American Research Products Inc Rat Protein S100 A9 S100A9 E
Catalog No. 5000328037
Encompass
Rat Protein S100-A9 (S100A9) ELISA kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28037 Supplier American Research Products Inc Supplier No. CSBEL020642RA
Rat Protein S100-A9 (S100A9) ELISA kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates Method Sandwich