Learn More
Biomatik Corporation Rat Protein Zero, Myelin (MPZ) ELISA Kit
Supplier:  Biomatik Corporation EKU06957
Encompass_Preferred
Rat Protein Zero, Myelin (MPZ) ELISA Kit for detecting MPZ in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06907. Gene ID: 24564. Target Synonyms: P0; DSS; CHM; CMT4E; CMT1; CMT1B; CMT2I; CMT2J; CMTDI3; HMSNIB; MPP; Myelin Protein Zero; Charcot-Marie-Tooth Neuropathy 1B; Myelin peripheral protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5154