Learn More
American Research Products Inc Rat prothrombin fragment 1 2
Catalog No. 5000329263
Encompass
Rat prothrombin fragment 1 2 F1 2 ELISA Kit from CUSABIO Detection Range 62 5 pmol/L-4000 pmol/L Sensitivity 15 6 pmol/L Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29263 Supplier American Research Products Inc Supplier No. CSBE13264R
Rat prothrombin fragment 1 2 F1 2 ELISA Kit from CUSABIO Detection Range 62 5 pmol/L-4000 pmol/L Sensitivity 15 6 pmol/L Sample serum plasma tissue homogenates Method Sandwich