Learn More
Biomatik Corporation Rat Protoporphyrinogen Oxidase (PPOX) ELISA Kit
Supplier:  Biomatik Corporation EKU06974
Encompass_Preferred
Rat Protoporphyrinogen Oxidase (PPOX) ELISA Kit for detecting PPOX in saliva, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZVN7. Gene ID: 289219. Target Synonyms: VP; PPO; V290M; Variegate Porphyria. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4648