Learn More
Cayman Chemical Rat Peroxisome Proliferator-Activated Receptor Alpha Reporter Assay System, 3x32 Well
SDP
Catalog No. 502971915
Encompass_Preferred
rPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00111-32, R00111]
Catalog No. 50-297-1915 Supplier Cayman Chemical Supplier No. 157643X32 WELL
rPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00111-32, R00111]