Learn More
Cayman Chemical Rat PRoxIsome ProlIfRatrA 96
SDP
Supplier:  Cayman Chemical 1576496 WELL
Encompass_Preferred
rPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00111-32, R00111]
Catalog No. 50-297-2568