Learn More
Cayman Chemical Rat Peroxisome Proliferator-Activated Receptor Delta Assay System, 96 Well
SDP
Catalog No. 502972202
Encompass_Preferred
rPPARδ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00121-32, R00121]
Catalog No. 50-297-2202 Supplier Cayman Chemical Supplier No. 1576596 WELL
rPPARδ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00121-32, R00121]