Learn More
Biomatik Corporation Rat P-Selectin Glycoprotein Ligand 1 (PSGL1) ELISA Kit
Supplier:  Biomatik Corporation EKU06976
Encompass_Preferred
Rat P-Selectin Glycoprotein Ligand 1 (PSGL1) ELISA Kit for detecting PSGL1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.50-4000pg/ml. Sensitivity: 28.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K5B0. Gene ID: 363930. Target Synonyms: CD162; SELPLG; CLA; Selectin P Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7695