Learn More
ACROBiosystems ACROBiosystems Rat PSMA / FOLH1 Protein, His Tag
Catalog No. 502106497
Encompass_Preferred
Recombinant Protein;1MG;Rat PSMA, His Tag (PSA-R5245) is expressed from human 293 cells (HEK293). It contains AA Lys 45 - Asp 752 (Accession # P70627-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 kDa.
Catalog No. 50-210-6497 Supplier ACROBiosystems Supplier No. PSAR52451MG
Recombinant Protein;1MG;Rat PSMA, His Tag (PSA-R5245) is expressed from human 293 cells (HEK293). It contains AA Lys 45 - Asp 752 (Accession # P70627-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 kDa.