Learn More
Biomatik Corporation Rat Pth (PaRathyroid hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF58603
Encompass_Preferred
Rat Pth (PaRathyroid hormone) ELISA Kit for detecting Pth in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-800pg/ml. Sensitivity: 7.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04089. Target Synonyms: Pth; PaRathyroid hormone; PaRathormone; PaRathyrin; PaRathormone; paRathormone; PaRathyrin; paRathyrin; paRathyroid hormone; paRathyroid hormone 1; PTH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5504