Learn More
American Research Products Inc Rat Pulmonary Surfactant assoc
Catalog No. 5000329746
Encompass
Rat Pulmonary Surfactant-associated protein B (SP-B) ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29746 Supplier American Research Products Inc Supplier No. CSBE12630R
Rat Pulmonary Surfactant-associated protein B (SP-B) ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich