Learn More
American Research Products Inc Rat Pulmonary Surfactant assoc
Supplier:  American Research Products Inc CSBE12631R
Encompass
Rat Pulmonary Surfactant-associated protein C (SP-C) ELISA kit from CUSABIO Detection Range 4 69 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29778