Learn More
American Research Products Inc Rat Pulmonary surfactant assoc
Catalog No. 5000326188
Encompass
Rat Pulmonary surfactant-associated protein A SP-A ELISA Kit from CUSABIO Detection Range 31 25 pg/ml - 2000 pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26188 Supplier American Research Products Inc Supplier No. CSBE08684R
Rat Pulmonary surfactant-associated protein A SP-A ELISA Kit from CUSABIO Detection Range 31 25 pg/ml - 2000 pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich