Learn More
BIOHIPPO Rat PVALB Microsample
SDP
Catalog No. 5000313987
Encompass
Rat PVALB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PVALB in Serum plasma and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PVALB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PVALB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PVALB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13987 Supplier BIOHIPPO Supplier No. BHE15211137
Rat PVALB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PVALB in Serum plasma and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PVALB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PVALB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PVALB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color