Learn More
Crystal Chem Inc Rat PYY ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 81502
Encompass
The Rat PYY ELISA is a highly sensitive assay that is used to quantify levels of rat PYY in serum and plasma.Highlights: Kits use sample size of only 25 µL. High sensitivity of 0.15 ng/mL. Dynamic range of 0.15 - 12.5 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7891