Learn More
Sino Biological Anti-RAB7A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80629T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat RAB7A / Rab-7a (rR RAB7A / Rab-7a; Catalog#80629-R07E; NP_076440.1; Met1-Cys207). RAB7A / Rab-7a specific IgG was purified by Rat RAB7A / Rab-7a affinity chromatography.
Catalog No. 50-240-8252