Learn More
Raybiotech Inc Rat RANTES/CCL5 ELISA Kit
Supplier:  Raybiotech Inc ELRRANTES1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat RANTES.
Catalog No. 50-195-0302