Learn More
Cayman Chemical Rat RAR-related Orphan Receptor Gamma Reporter Assay System, 96 Well
SDP
Catalog No. 502913188
Encompass_Preferred
rRORγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R04001, R04001-32]
Catalog No. 50-291-3188 Supplier Cayman Chemical Supplier No. 3615496 WELL
rRORγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R04001, R04001-32]