Learn More
Sino Biological Recombinant Anti-RBP4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80008R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat RBP4 (rR RBP4; Catalog#80008-R08H; NP_037294.1; Met1-Leu201).
Catalog No. 50-241-2403