Learn More
STEMCELL Technologies Rat Recombinant GM-CSF, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78018
Encompass
Use granulocyte-macrophage colony-stimulating factor (GM-CSF) to induce the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages (Barreda et al. Dev Comp Immunol, 2004). GM-CSF can stimulate the development of DCs that ingest, process, and present antigens to the immune system (Francisco-Cruz et al. Med Oncol, 2014). It is produced by multiple cell types, including macrophages, dendritic cells, endothelial cells, and fibroblasts. Together with other cytokines, GM-CSF also stimulates erythroid and megakaryocyte progenitor cells (Barreda et al.). Recombinant rat GM-CSF is reactive with mouse cells (Oaks et al. J Interferon Cytokine Res, 1995). Rat recombinant GM-CSF from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 20 pg/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: Colony-stimulating factor 2, CSF-2, MGI-1GM, Pluripoitin-alpha. Visit STEMCELL.com for more information.
Catalog No. 50-197-6279
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov