Learn More
Sino Biological Rat REG3A Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80258R08H100
Encompass_Preferred
A DNA sequence encoding the rat REG3A (BAA04904.1) (Met1-Gln174) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3383