Learn More
American Research Products Inc Rat Regenerating islet deriVed
Catalog No. 5000327201
Encompass
Rat Regenerating islet-deriVed protein 3-gamma (REG3G) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27201 Supplier American Research Products Inc Supplier No. CSBEL019549RA
Rat Regenerating islet-deriVed protein 3-gamma (REG3G) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich