Learn More
American Research Products Inc Rat regulated on actiVation in
Catalog No. 5000329608
Encompass
Rat regulated on actiVation in normal T-cell eXpressed and secreted/C-C motif chemokine 5 (RANTES/CCL5) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29608 Supplier American Research Products Inc Supplier No. CSBE07398R
Rat regulated on actiVation in normal T-cell eXpressed and secreted/C-C motif chemokine 5 (RANTES/CCL5) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich