Learn More
American Research Products Inc Rat regulated on actiVation in
Supplier:  American Research Products Inc CSBE07398RIS
Encompass
Rat regulated on actiVation in normal T-cell eXpressed and secreted/C-C motif chemokine 5 (RANTES/CCL5) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29609