Learn More
Biomatik Corporation Rat Regulator Of G Protein Signaling 6 (RGS6) ELISA Kit
Supplier:  Biomatik Corporation EKU07058
Encompass_Preferred
Rat Regulator Of G Protein Signaling 6 (RGS6) ELISA Kit for detecting RGS6 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1LS67. Gene ID: 54295. Target Synonyms: GAP; S914. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8782