Learn More
Biomatik Corporation Rat Regulator Of G Protein Signaling 9 (RGS9) ELISA Kit
Supplier:  Biomatik Corporation EKU11395
Encompass_Preferred
Rat Regulator Of G Protein Signaling 9 (RGS9) ELISA Kit for detecting RGS9 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49805. Gene ID: 29481. Target Synonyms: PERRS; RGS9L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5388