Learn More
Raybiotech Inc Rat Relative Mitochondrial DNA Copy Number Quantification Kit
Catalog No. NC3909503
Encompass_Preferred
The RayBio® Rat Relative Mitochondrial DNA Copy Number Quantification Kit is a Taqman™ probe-based qPCR assay for the specific quantitative measurement of rat mtDNA-CN.
Catalog No. NC3909503 Supplier Raybiotech Inc Supplier No. MTRCNQ100
The RayBio® Rat Relative Mitochondrial DNA Copy Number Quantification Kit is a Taqman™ probe-based qPCR assay for the specific quantitative measurement of rat mtDNA-CN.