Learn More
Biomatik Corporation Rat Relaxin 3 (RLN3) ELISA Kit
Supplier:  Biomatik Corporation EKU07067
Encompass_Preferred
Rat Relaxin 3 (RLN3) ELISA Kit for detecting RLN3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.46-10000pg/ml. Sensitivity: 43.6pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8BFS3. Gene ID: 266997. Target Synonyms: ZINS4; RXN3; H3; INSL7; Insulin-like peptide 7; Prorelaxin H3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8780