Learn More
Biomatik Corporation Rat Renal Tumor Antigen (RAGE) ELISA Kit
Supplier:  Biomatik Corporation EKU07070
Encompass_Preferred
Rat Renal Tumor Antigen (RAGE) ELISA Kit for detecting RAGE in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6AXN5. Gene ID: 362787. Target Synonyms: MOK; RAGE1; MAPK/MAK/MRK Overlapping Kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5382