Learn More
American Research Products Inc Rat Renalase Monoamine OXidas
Catalog No. 5000327422
Encompass
Rat Renalase/ Monoamine OXidase-C (RNLS/ MAO-C) ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27422 Supplier American Research Products Inc Supplier No. CSBEL002965RA
Rat Renalase/ Monoamine OXidase-C (RNLS/ MAO-C) ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Sandwich