Learn More
Invitrogen™ Rat Renin-1 ELISA Kit
Description
Rat Renin-1 quantitates rat Renin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Renin-1.
Specifications
Specifications
| Accession Number | P06281, P08424 |
| Assay Range | 27.4-20000 pg/mL |
| Assay Sensitivity | 30 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse, Rat |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnrangiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.