Learn More
CPC Scientific Ac-His-Pro-Phe-Val-Sta-Leu-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific RENI011A
Encompass
A specific rat renin inhibitor.ONE-LETTER SEQUENCE: Ac-HPFV-(Sta)-LF-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:957.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: CardiovascularREFERENCES: Iyer, SN. et al. J. Cardiovasc. Pharmacol. 31, 464 (1998)
Catalog No. 50-211-0367