Learn More
Braintree Scientific Tailveiner Restrainer Rat, up to 120gms, 1 3/4"""" ID
SDP
Catalog No. NC9596898
Encompass
Tail vein injection made easier! The Tailveiner is the first easy-to-use mouse and rat restrainer specifically designed for tail vein injections. Slotted tube permits the rat or mouse to be quickly pulled in by the tail. The animal is then restrained by a plug which slides easily to accommodate any size rat or mouse, but locks automatically when handle is released. After dosing, the handle is lifted and the plug is easily removed. The Tailveiner is the fastest restrainer yet for intravenous injections in small animals. **Sold for research purposes only and not for human use**
Catalog No. NC9596898 Supplier Braintree Scientific Supplier No. RTV170
Tail vein injection made easier! The Tailveiner is the first easy-to-use mouse and rat restrainer specifically designed for tail vein injections. Slotted tube permits the rat or mouse to be quickly pulled in by the tail. The animal is then restrained by a plug which slides easily to accommodate any size rat or mouse, but locks automatically when handle is released. After dosing, the handle is lifted and the plug is easily removed. The Tailveiner is the fastest restrainer yet for intravenous injections in small animals. **Sold for research purposes only and not for human use**