Learn More
Braintree Scientific Flat Bottom Restrainer, 1.5"" dia x 4"", 15-30gm Mice
SDP
Catalog No. NC9398939
Encompass
Economy Flat Bottomed Restrainers. Individual semi cylindrical plastic restrainers in small, medium and large. Each unit has an open slot on the bottom (for intra-peritoneal injections), a hole on the top (for the neck), and a slotted tail piece (for intravenous injections). Front slots provide ample ventilation. **Sold for research purposes only and not for human use**
Catalog No. NC9398939 Supplier Braintree Scientific Supplier No. FBS
 Restricted Item
Add to Cart
Add to Cart
Economy Flat Bottomed Restrainers. Individual semi cylindrical plastic restrainers in small, medium and large. Each unit has an open slot on the bottom (for intra-peritoneal injections), a hole on the top (for the neck), and a slotted tail piece (for intravenous injections). Front slots provide ample ventilation. **Sold for research purposes only and not for human use**