Learn More
Biomatik Corporation Rat Retinoic Acid Receptor Alpha (RARa) ELISA Kit
Supplier:  Biomatik Corporation EKU07094
Encompass_Preferred
Rat Retinoic Acid Receptor Alpha (RARa) ELISA Kit for detecting RARa in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q499N1. Gene ID: 24705. Target Synonyms: RAR-A; NR1B1; RAR; RAR-Alpha; Nuclear Receptor Subfamily 1 Group B Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4697