Learn More
American Research Products Inc Rat Rheumatoid Factor RF ELI
Catalog No. 5000329466
Encompass
Rat Rheumatoid Factor (RF) ELISA Kit from CUSABIO Detection Range 15 6 mIU/mL-1000 mIU/mL Sensitivity 3 9 mIU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29466 Supplier American Research Products Inc Supplier No. CSBE13666R
Rat Rheumatoid Factor (RF) ELISA Kit from CUSABIO Detection Range 15 6 mIU/mL-1000 mIU/mL Sensitivity 3 9 mIU/mL Sample serum plasma Method Sandwich