Learn More
Biomatik Corporation Rat Ribonuclease T2 (RNASET2) CLIA Kit
Supplier:  Biomatik Corporation EKU09757
Encompass_Preferred
Rat Ribonuclease T2 (RNASET2) CLIA Kit for detecting RNASET2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: B5DF79. Target Synonyms: Rnase-T2; RNASE6PL; RNASE6-PL; Ribonuclease 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4470