Learn More
BIOHIPPO Rat RLN Microsample
SDP
Catalog No. 5000315108
Encompass
Rat RLN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN in serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 9 05 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat RLN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat RLN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat RLN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15108 Supplier BIOHIPPO Supplier No. BHE15210878
Rat RLN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN in serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 9 05 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat RLN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat RLN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat RLN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color