Learn More
BIOHIPPO Rat RLN3 Microsample
SDP
Catalog No. 5000314354
Encompass
Rat RLN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN3 in Serum plasma and other biological fluids It provides a detection range of 156 25-10000 pg/mL with a sensitivity of 44 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat RLN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat RLN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat RLN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14354 Supplier BIOHIPPO Supplier No. BHE15210855
Rat RLN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN3 in Serum plasma and other biological fluids It provides a detection range of 156 25-10000 pg/mL with a sensitivity of 44 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat RLN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat RLN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat RLN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color