Learn More
BIOHIPPO Rat RNLS Microsample
SDP
Catalog No. 5000314461
Encompass
Rat RNLS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RNLS in serum plasma and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 69 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat RNLS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat RNLS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat RNLS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14461 Supplier BIOHIPPO Supplier No. BHE15210947
Rat RNLS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RNLS in serum plasma and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 69 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat RNLS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat RNLS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat RNLS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color