Learn More
American Research Products Inc Rat rudimental boVine serum al
Catalog No. 5000329493
Encompass
Rat rudimental boVine serum albumin (BSA) check-up ELISA kit from CUSABIO Detection Range 0 312 g/mL-80 g/mL Sensitivity 0 312 g/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29493 Supplier American Research Products Inc Supplier No. CSBE08634R
Rat rudimental boVine serum albumin (BSA) check-up ELISA kit from CUSABIO Detection Range 0 312 g/mL-80 g/mL Sensitivity 0 312 g/mL Sample serum plasma tissue homogenates Method Competitive