Learn More
Biomatik Corporation Rat S100 Calcium Binding Protein A9 (S100A9) ELISA Kit
Supplier:  Biomatik Corporation EKU09085
Encompass_Preferred
Rat S100 Calcium Binding Protein A9 (S100A9) ELISA Kit for detecting S100A9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50116. Gene ID: 94195. Target Synonyms: MRP14; MIF; 60B8AG; CFAG; NIF; CAGB; CGLB; L1AG; P14; Calgranulin B; Migration inhibitory factor-related protein 14; Calprotectin L1H subunit; Leukocyte L1 complex heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4510